Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRPB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115068.5 | NP_190777.1 | 3 - 4 | 326 | 96 | |
GenBank mRNA | BT005984.1 | - | 3 - 4 | 224 | 96 | |
U28048.1 | - | 3 - 4 | 275 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |