Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCS15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339560.1 | NP_001327019.1 | 1 - 2 | 1053 | 67 | |
NM_115076.4 | NP_190784.1 | 2 - 3 | 971 | 67 | ||
GenBank mRNA | DQ446759.1 | - | 2 - 3 | 894 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |