Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTSZ2-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339586.1 | NP_001327517.1 | 5 - 6 | 1489 | 72 | |
NM_001339587.1 | NP_001327516.1 | 6 - 7 | 1460 | 72 | ||
NM_115135.3 | NP_190843.1 | 5 - 6 | 1392 | 72 | ||
GenBank mRNA | AF384167.1 | - | 4 - 5 | 1134 | 72 | |
AY150504.1 | - | 4 - 5 | 1134 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |