Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAP22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115142.5 | NP_190850.2 | 4 - 5 | 1204 | 66 | |
GenBank mRNA | AF492668.1 | - | 4 - 5 | 1001 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |