Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339590.1 | NP_001325739.1 | 9 - 10 | 1164 | 112 | |
NM_115144.4 | NP_190852.2 | 9 - 10 | 1126 | 112 | ||
GenBank mRNA | AF367327.1 | - | 9 - 10 | 1058 | 112 | |
BT000511.1 | - | 9 - 10 | 969 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |