Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115145.2 | NP_190853.1 | 9 - 10 | 1777 | 64 | |
GenBank mRNA | U86700.1 | - | 9 - 10 | 1696 | 64 | |
Y07917.1 | - | 1706 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |