Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115154.3 | NP_566975.1 | 3 - 4 | 454 | 119 | |
NM_202691.1 | NP_974420.1 | 3 - 4 | 409 | 119 | ||
GenBank mRNA | AF256535.1 | - | 3 - 4 | 362 | 119 | |
AK227189.1 | - | 3 - 4 | 320 | 119 | ||
AY064964.1 | - | 3 - 4 | 422 | 119 | ||
AY133650.1 | - | 3 - 4 | 238 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |