Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035773.2 | NP_001030850.1 | 5 - 6 | 2006 | 130 | |
NM_115169.3 | NP_190877.1 | 5 - 6 | 2009 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |