Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CP29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203150.1 | NP_001190079.1 | 4 - 5 | 1236 | 80 | |
NM_115206.3 | NP_190914.1 | 4 - 5 | 1159 | 80 | ||
GenBank mRNA | AY039909.1 | - | 4 - 5 | 1173 | 80 | |
AY087840.1 | - | 4 - 5 | 1159 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |