Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115222.3 | NP_190930.1 | 2 - 3 | 419 | 78 | |
GenBank mRNA | AK226578.1 | - | 2 - 3 | 303 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |