Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115222.3 | NP_190930.1 | 6 - 7 | 682 | 84 | |
GenBank mRNA | AK226578.1 | - | 6 - 7 | 566 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |