Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FIP37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115277.5 | NP_190985.1 | 11 - 12 | 1021 | 138 | |
GenBank mRNA | AF084570.1 | - | 11 - 12 | 903 | 138 | |
AY063940.1 | - | 11 - 12 | 928 | 138 | ||
AY113943.1 | - | 11 - 12 | 837 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |