Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115282.4 | NP_190990.1 | 1 - 2 | 2088 | 79 | |
GenBank mRNA | AY056315.1 | - | 1 - 2 | 1711 | 79 | |
AY080840.1 | - | 1 - 2 | 1728 | 79 | ||
AY113991.1 | - | 1 - 2 | 1525 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |