Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203156.1 | NP_001190085.1 | 4 - 5 | 505 | 64 | |
NM_115288.8 | NP_190996.3 | 4 - 5 | 629 | 64 | ||
GenBank mRNA | AB440793.1 | - | 3 - 4 | 367 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |