Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035780.3 | NP_001030857.1 | 6 - 7 | 921 | 92 | |
NM_001339658.1 | NP_001319744.1 | 4 - 5 | 337 | 92 | ||
NM_001339659.1 | NP_001325502.1 | 5 - 6 | 1050 | 92 | ||
NM_001339660.1 | NP_001325503.1 | 4 - 5 | 432 | 92 | ||
NM_115292.4 | NP_191000.3 | 6 - 7 | 725 | 92 | ||
GenBank mRNA | AY254038.2 | - | 6 - 7 | 679 | 92 | |
AY885245.1 | - | 6 - 7 | 600 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |