Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115294.6 | NP_191002.1 | 3 - 4 | 548 | 90 | |
GenBank mRNA | AY070397.1 | - | 3 - 4 | 351 | 90 | |
AY142590.1 | - | 3 - 4 | 321 | 90 | ||
M86357.1 | - | 3 - 4 | 379 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |