Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBL17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115322.4 | NP_567005.2 | 5 - 6 | 1491 | 81 | |
GenBank mRNA | AF462827.1 | - | 5 - 6 | 1185 | 81 | |
AY142006.1 | - | 5 - 6 | 1159 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |