Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115329.3 | NP_567007.1 | 6 - 7 | 1711 | 124 | |
GenBank mRNA | BT046200.1 | - | 6 - 7 | 1610 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |