Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339702.1 | NP_001326426.1 | 20 - 21 | 3148 | 66 | |
NM_001339703.1 | NP_001326427.1 | 19 - 20 | 3254 | 66 | ||
NM_115344.4 | NP_191047.3 | 19 - 20 | 3169 | 66 | ||
GenBank mRNA | AB290928.1 | - | 20 - 21 | 3037 | 66 | |
EU257694.1 | - | 20 - 21 | 3022 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |