Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TON1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115358.5 | NP_567013.1 | 4 - 5 | 673 | 78 | |
GenBank mRNA | AY075696.1 | - | 4 - 5 | 565 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |