Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115359.3 | NP_191061.1 | 2 - 3 | 391 | 69 | |
NM_202708.2 | NP_974437.1 | 1 - 2 | 508 | 69 | ||
GenBank mRNA | AY060585.1 | - | 2 - 3 | 369 | 69 | |
AY142054.1 | - | 1 - 2 | 295 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |