Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115359.3 | NP_191061.1 | 6 - 7 | 835 | 79 | |
NM_202708.2 | NP_974437.1 | 5 - 6 | 952 | 79 | ||
GenBank mRNA | AY060585.1 | - | 6 - 7 | 813 | 79 | |
AY142054.1 | - | 5 - 6 | 739 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |