Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115361.3 | NP_191063.1 | 3 - 4 | 590 | 95 | |
GenBank mRNA | AF370186.1 | - | 3 - 4 | 427 | 95 | |
AY059144.1 | - | 3 - 4 | 327 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |