Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEXO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115384.4 | NP_567017.2 | 14 - 15 | 1667 | 80 | |
GenBank mRNA | AM493720.1 | - | 14 - 15 | 1446 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |