Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115390.3 | NP_191092.1 | 10 - 11 | 3136 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |