Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OVA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084826.2 | NP_001078295.1 | 9 - 10 | 1893 | 87 | |
NM_115398.3 | NP_191100.1 | 10 - 11 | 1812 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |