Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDIR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115410.3 | NP_191112.1 | 5 - 6 | 578 | 98 | |
GenBank mRNA | AF386995.1 | - | 5 - 6 | 548 | 98 | |
AK317055.1 | - | 5 - 6 | 565 | 98 | ||
BT006305.1 | - | 4 - 5 | 410 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |