Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339748.1 | NP_001319763.1 | 10 - 11 | 1837 | 65 | |
NM_001339749.1 | NP_001326016.1 | 9 - 10 | 2026 | 65 | ||
NM_001339750.1 | NP_001326018.1 | 9 - 10 | 2026 | 65 | ||
NM_001339751.1 | NP_001326017.1 | 11 - 12 | 1315 | 65 | ||
NM_115443.4 | NP_567027.2 | 11 - 12 | 1321 | 65 | ||
NM_202716.3 | NP_974445.2 | 12 - 13 | 1534 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |