Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSLA14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115458.5 | NP_191159.2 | 2 - 3 | 613 | 95 | |
GenBank mRNA | KJ138786.1 | - | 2 - 3 | 412 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |