Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRLK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339765.1 | NP_001319765.1 | 1 - 2 | 2031 | 69 | |
NM_001339766.1 | NP_001325935.1 | 2 - 3 | 1617 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |