Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339805.1 | NP_001326776.1 | 5 - 6 | 963 | 126 | |
NM_115529.2 | NP_191229.1 | 5 - 6 | 900 | 126 | ||
GenBank mRNA | DQ056628.1 | - | 5 - 6 | 803 | 126 | |
EU280151.1 | - | 5 - 6 | 866 | 126 | ||
EU280152.1 | - | 5 - 6 | 803 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |