Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339806.1 | NP_001325423.1 | 2 | 780 | 60 | |
NM_115530.2 | NP_191230.1 | 1 | 867 | 60 | ||
GenBank mRNA | AF224762.1 | - | 1 | 324 | 60 | |
AY042831.1 | - | 1 | 368 | 60 | ||
BT002607.1 | - | 1 | 311 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |