Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSRP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115550.4 | NP_191250.1 | 1 - 2 | 192 | 61 | |
GenBank mRNA | BT014859.1 | - | 1 - 2 | 93 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |