Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIP5K4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339834.1 | NP_001319773.1 | 8 - 9 | 2933 | 74 | |
NM_115555.2 | NP_191255.1 | 7 - 8 | 2986 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |