Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIGYIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339841.1 | NP_001327069.1 | 1 - 2 | 321 | 68 | |
NM_115568.4 | NP_567044.1 | 1 - 2 | 321 | 68 | ||
GenBank mRNA | AB195717.1 | - | 1 - 2 | 90 | 68 | |
AY050354.1 | - | 1 - 2 | 187 | 68 | ||
AY087545.1 | - | 1 - 2 | 187 | 68 | ||
AY097397.1 | - | 1 - 2 | 90 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |