Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339858.1 | NP_001319779.1 | 24 - 25 | 2996 | 112 | |
NM_001339860.1 | NP_001325645.1 | 24 - 25 | 3761 | 112 | ||
NM_001339861.1 | NP_001325651.1 | 24 - 25 | 3815 | 112 | ||
NM_001339862.1 | NP_001325648.1 | 24 - 25 | 3430 | 112 | ||
NM_001339863.1 | NP_001325647.1 | 24 - 25 | 3468 | 112 | ||
NM_001339864.1 | NP_001325650.1 | 24 - 25 | 3137 | 112 | ||
NM_001339865.1 | NP_001325649.1 | 23 - 24 | 3132 | 112 | ||
NM_001339866.1 | NP_001325646.1 | 23 - 24 | 3170 | 112 | ||
NM_115601.3 | NP_567048.1 | 24 - 25 | 3034 | 112 | ||
GenBank mRNA | AK318626.1 | - | 23 - 24 | 2987 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |