Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ILP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120941.2 | NP_196472.1 | 2 - 3 | 1279 | 134 | |
GenBank mRNA | AB253763.1 | - | 2 - 3 | 1279 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Arabidopsis thaliana Inventoried Probe spans exons |