Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTHSC70-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120996.4 | NP_196521.1 | 5 - 6 | 1696 | 72 | |
GenBank mRNA | AF217458.1 | - | 5 - 6 | 1469 | 72 | |
BT046197.1 | - | 5 - 6 | 1411 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |