Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDH3-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085086.1 | NP_001078555.1 | 4 - 5 | 393 | 73 | |
NM_001203339.2 | NP_001190268.1 | 4 - 5 | 503 | 73 | ||
NM_120997.4 | NP_196522.1 | 3 - 4 | 332 | 73 | ||
GenBank mRNA | AY070732.1 | - | 3 - 4 | 303 | 73 | |
AY074861.1 | - | 3 - 4 | 283 | 73 | ||
BT010154.1 | - | 3 - 4 | 219 | 73 |
Gene Symbol: SDH3-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119373.3 | NP_194948.2 | 3 - 4 | 345 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |