Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCME | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121591.5 | NP_197090.2 | 10 - 11 | 1618 | 94 | |
NM_203057.1 | NP_974786.1 | 7 - 8 | 1380 | 94 | ||
GenBank mRNA | AY045829.1 | - | 10 - 11 | 1239 | 94 | |
AY091367.1 | - | 10 - 11 | 1120 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |