Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344487.1 | NP_001331886.1 | 6 - 7 | 925 | 93 | |
NM_001344488.1 | NP_001331887.1 | 6 - 7 | 767 | 93 | ||
NM_123716.3 | NP_199163.1 | 6 - 7 | 879 | 93 | ||
NM_203147.2 | NP_974876.1 | 6 - 7 | 925 | 93 | ||
GenBank mRNA | AY057568.1 | - | 6 - 7 | 694 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |