Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRMT4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124279.5 | NP_199713.2 | 14 - 15 | 1663 | 106 | |
NM_203184.2 | NP_974913.1 | 14 - 15 | 1597 | 106 | ||
GenBank mRNA | BT030319.1 | - | 14 - 15 | 1513 | 106 | |
BT030323.1 | - | 14 - 15 | 1513 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |