Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124376.4 | NP_568717.2 | 9 - 10 | 1364 | 130 | |
NM_203189.2 | NP_974918.1 | 9 - 10 | 1232 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |