Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345181.1 | NP_001330467.1 | 4 - 5 | 843 | 95 | |
NM_125000.4 | NP_568836.2 | 5 - 6 | 722 | 95 | ||
NM_203211.1 | NP_974940.1 | 5 - 6 | 645 | 95 | ||
GenBank mRNA | AF507916.1 | - | 5 - 6 | 615 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |