Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037031.1 | NP_001032108.1 | 14 - 15 | 1440 | 84 | |
NM_001037032.2 | NP_001032109.2 | 14 - 15 | 1440 | 84 | ||
NM_001125993.1 | NP_001119465.1 | 14 - 15 | 1440 | 84 | ||
NM_001345415.1 | NP_001332003.1 | 14 - 15 | 1470 | 84 | ||
NM_125434.4 | NP_200849.2 | 14 - 15 | 1626 | 84 | ||
NM_203240.3 | NP_974969.1 | 14 - 15 | 1569 | 84 | ||
GenBank mRNA | AK175813.1 | - | 14 - 15 | 1440 | 84 | |
AK220973.1 | - | 14 - 15 | 1440 | 84 | ||
AK221190.1 | - | 14 - 15 | 1440 | 84 | ||
AK221249.1 | - | 14 - 15 | 1440 | 84 | ||
AK221292.1 | - | 14 - 15 | 1440 | 84 | ||
AK316983.1 | - | 14 - 15 | 1440 | 84 | ||
AY139752.1 | - | 14 - 15 | 1439 | 84 | ||
AY700572.1 | - | 14 - 15 | 1441 | 84 | ||
BT004542.1 | - | 13 - 14 | 1202 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |