Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BON1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125583.4 | NP_568944.1 | 4 - 5 | 434 | 139 | |
NM_203248.2 | NP_974977.1 | 3 - 4 | 469 | 139 | ||
GenBank mRNA | AX380787.1 | - | 3 - 4 | 391 | 139 | |
AY062824.1 | - | 4 - 5 | 425 | 139 | ||
AY085339.1 | - | 4 - 5 | 426 | 139 | ||
BT010555.1 | - | 3 - 4 | 347 | 139 |
Gene Symbol: AT5G61910 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203661.1 | NP_001190590.1 | 10 - 11 | 2870 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |