Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198200.2 | NP_001185129.1 | 1 - 2 | 390 | 89 | |
NM_001333042.1 | NP_001319133.1 | 1 - 2 | 390 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |