Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333051.1 | NP_001319134.1 | 2 - 3 | 297 | 66 | |
NM_001333052.1 | NP_001321942.1 | 1 - 2 | 376 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |