Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP76C6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103092.3 | NP_174633.1 | 1 - 2 | 415 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |