Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IP5PI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333086.1 | NP_001322580.1 | 2 - 3 | 246 | 130 | |
NM_103135.4 | NP_564437.1 | 2 - 3 | 307 | 130 | ||
NM_179414.1 | NP_849745.1 | 2 - 3 | 229 | 130 | ||
NM_202231.1 | NP_973960.1 | 2 - 3 | 229 | 130 | ||
GenBank mRNA | AK227368.1 | - | 2 - 3 | 197 | 130 | |
BT006448.1 | - | 2 - 3 | 176 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |